The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099855.1 | NP_001093325.1 | 15 - 16 | 1831 | 112 |
NM_001171876.1 | NP_001165347.1 | 16 - 17 | 1879 | 112 | |
NM_001171877.1 | NP_001165348.1 | 11 - 12 | 1696 | 112 | |
NM_001171878.1 | NP_001165349.1 | 12 - 13 | 1813 | 112 | |
NM_001171879.1 | NP_001165350.1 | 13 - 14 | 1861 | 112 | |
NM_005369.4 | NP_005360.3 | 12 - 13 | 1813 | 112 | |
XM_005262413.4 | XP_005262470.1 | 17 - 18 | 2027 | 112 | |
XM_011531339.2 | XP_011529641.1 | 17 - 18 | 2086 | 112 | |
XM_017029529.1 | XP_016885018.1 | 16 - 17 | 1848 | 112 | |
XM_017029530.1 | XP_016885019.1 | 16 - 17 | 1848 | 112 | |
XM_017029531.1 | XP_016885020.1 | 16 - 17 | 1781 | 112 | |
XM_017029532.1 | XP_016885021.1 | 13 - 14 | 1938 | 112 | |
GenBank mRNA | AB085901.1 | - | 12 - 13 | 1490 | 112 |
AB085902.1 | - | 13 - 14 | 1538 | 112 | |
AK296488.1 | - | 11 - 12 | 1696 | 112 | |
AK296529.1 | - | 15 - 16 | 1931 | 112 | |
AK302957.1 | - | 17 - 18 | 1827 | 112 | |
AK313900.1 | - | 12 - 13 | 1673 | 112 | |
AK316416.1 | - | 16 - 17 | 1867 | 112 | |
AL117234.1 | - | 14 - 15 | 1714 | 112 | |
X12556.1 | - | 12 - 13 | 1664 | 112 | |
X13230.1 | - | - | 418 | 112 |