The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099855.1 | NP_001093325.1 | 18 - 19 | 2090 | 116 |
NM_001171876.1 | NP_001165347.1 | 19 - 20 | 2138 | 116 | |
NM_001171877.1 | NP_001165348.1 | 14 - 15 | 1955 | 116 | |
NM_001171878.1 | NP_001165349.1 | 15 - 16 | 2072 | 116 | |
NM_001171879.1 | NP_001165350.1 | 16 - 17 | 2120 | 116 | |
NM_005369.4 | NP_005360.3 | 15 - 16 | 2072 | 116 | |
XM_005262413.4 | XP_005262470.1 | 20 - 21 | 2286 | 116 | |
XM_011531339.2 | XP_011529641.1 | 20 - 21 | 2345 | 116 | |
XM_017029529.1 | XP_016885018.1 | 19 - 20 | 2107 | 116 | |
XM_017029530.1 | XP_016885019.1 | 19 - 20 | 2107 | 116 | |
XM_017029531.1 | XP_016885020.1 | 19 - 20 | 2040 | 116 | |
XM_017029532.1 | XP_016885021.1 | 16 - 17 | 2197 | 116 | |
GenBank mRNA | AB085901.1 | - | 15 - 16 | 1749 | 116 |
AB085902.1 | - | 16 - 17 | 1797 | 116 | |
AK296488.1 | - | 14 - 15 | 1955 | 116 | |
AK296529.1 | - | 18 - 19 | 2190 | 116 | |
AK302957.1 | - | 20 - 21 | 2086 | 116 | |
AK313900.1 | - | 15 - 16 | 1932 | 116 | |
AK316416.1 | - | 19 - 20 | 2126 | 116 | |
AL117234.1 | - | 17 - 18 | 1973 | 116 | |
J03639.1 | - | - | 805 | 116 | |
X12556.1 | - | 15 - 16 | 1923 | 116 | |
X13230.1 | - | - | 677 | 116 |