The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286746.1 | NP_001273675.1 | 7 - 8 | 888 | 91 |
NM_001286747.1 | NP_001273676.1 | 6 - 7 | 683 | 91 | |
NM_024808.3 | NP_079084.3 | 7 - 8 | 790 | 91 | |
XM_006719868.3 | XP_006719931.2 | 5 - 6 | 647 | 91 | |
GenBank mRNA | AK026277.1 | - | 2 - 3 | 115 | 91 |
AK291496.1 | - | 7 - 8 | 616 | 91 | |
AK298608.1 | - | 6 - 7 | 509 | 91 | |
AK300758.1 | - | 6 - 7 | 588 | 91 | |
BC025367.2 | - | 7 - 8 | 601 | 91 | |
BC044213.1 | - | 6 - 7 | 414 | 91 | |
BC056876.1 | - | 7 - 8 | 613 | 91 | |
BX648605.1 | - | 7 - 8 | 888 | 91 | |
EU834129.1 | - | 7 - 8 | 696 | 91 |