The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286746.1 | NP_001273675.1 | 8 - 9 | 1113 | 111 |
NM_001286747.1 | NP_001273676.1 | 7 - 8 | 908 | 111 | |
NM_024808.3 | NP_079084.3 | 8 - 9 | 1015 | 111 | |
XM_006719868.3 | XP_006719931.2 | 6 - 7 | 872 | 111 | |
GenBank mRNA | AK026277.1 | - | 3 - 4 | 340 | 111 |
AK291496.1 | - | 8 - 9 | 841 | 111 | |
AK298608.1 | - | 7 - 8 | 734 | 111 | |
AK300758.1 | - | 7 - 8 | 813 | 111 | |
BC025367.2 | - | 8 - 9 | 826 | 111 | |
BC044213.1 | - | 7 - 8 | 639 | 111 | |
BC056876.1 | - | 8 - 9 | 838 | 111 | |
EU834129.1 | - | 8 - 9 | 921 | 111 |