Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- DIO3OS
- Entrez Gene ID
-
64150
- Gene Name
- DIO3 opposite strand/antisense RNA (head to head)
- Gene Aliases
-
C14orf134, DIO3-AS1, DIO3-OS, NCRNA00041
- Chromosome Location
-
Chr.14: 101552223 - 101555676 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC065701.1 |
- |
1 - 2 |
127 |
109 |
View Full Product Details