The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017923.1 | NP_001017923.1 | 2 - 3 | 793 | 70 |
GenBank mRNA | AF250395.1 | - | 2 - 3 | 777 | 70 |
AK126985.1 | - | 1 - 2 | 549 | 70 | |
BC030258.1 | - | 1 - 2 | 531 | 70 | |
BC105030.1 | - | 2 - 3 | 786 | 70 | |
BC105032.1 | - | 2 - 3 | 786 | 70 | |
BU624091.1 | - | 3 - 4 | 70 |