Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMC2
Entrez Gene ID
10592
Gene Name
structural maintenance of chromosomes 2
Gene Aliases
CAP-E, CAPE, SMC-2, SMC2L1
Chromosome Location
Chr.9: 104093808 - 104141419 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042550.1 NP_001036015.1 9 - 10 1364 97
NM_001042551.1 NP_001036016.1 9 - 10 1302 97
NM_001265602.1 NP_001252531.1 9 - 10 1318 97
NM_006444.2 NP_006435.2 9 - 10 1293 97
XM_006716933.3 XP_006716996.1 9 - 10 1215 97
XM_011518148.2 XP_011516450.1 9 - 10 1178 97
XM_011518149.2 XP_011516451.1 9 - 10 1220 97
XM_011518150.2 XP_011516452.1 8 - 9 1082 97
XM_011518151.2 XP_011516453.1 8 - 9 1060 97
XM_011518152.2 XP_011516454.1 9 - 10 1244 97
XM_011518153.1 XP_011516455.1 9 - 10 1244 97
XM_017014206.1 XP_016869695.1 9 - 10 1364 97
XM_017014207.1 XP_016869696.1 9 - 10 1266 97
XM_017014208.1 XP_016869697.1 9 - 10 1306 97
XM_017014209.1 XP_016869698.1 8 - 9 1193 97
XM_017014210.1 XP_016869699.1 8 - 9 1083 97
XM_017014211.1 XP_016869700.1 8 - 9 1172 97
XM_017014212.1 XP_016869701.1 8 - 9 1039 97
GenBank mRNA AF092563.1 - 9 - 10 1228 97
AK001485.1 - 9 - 10 1350 97
AK292599.1 - 9 - 10 1241 97
AK307752.1 - 8 - 9 1061 97
AL833191.1 - 9 - 10 1302 97
AY739716.1 - 9 - 10 1194 97
BC032705.1 - 9 - 10 1351 97
BC130385.1 - 9 - 10 1148 97
BC144163.1 - 9 - 10 1164 97

View Full Product Details