The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003045.4 | NP_003036.1 | 9 - 10 | 1682 | 75 |
XM_005266507.3 | XP_005266564.1 | 9 - 10 | 1477 | 75 | |
XM_017020713.1 | XP_016876202.1 | 9 - 10 | 1682 | 75 | |
XM_017020714.1 | XP_016876203.1 | 9 - 10 | 1621 | 75 | |
XM_017020715.1 | XP_016876204.1 | 9 - 10 | 1503 | 75 | |
XM_017020716.1 | XP_016876205.1 | 9 - 10 | 1693 | 75 | |
GenBank mRNA | AB105377.1 | - | 9 - 10 | 1569 | 75 |
AF078107.1 | - | 8 - 9 | 1415 | 75 | |
AK312818.1 | - | 9 - 10 | 1456 | 75 | |
BC063303.1 | - | 9 - 10 | 1479 | 75 | |
BC069358.1 | - | 8 - 9 | 1364 | 75 | |
BC115407.1 | - | 8 - 9 | 1364 | 75 | |
X57303.1 | - | 9 - 10 | 1445 | 75 | |
X59155.1 | - | 9 - 10 | 1442 | 75 |