Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
JPH3
Entrez Gene ID
57338
Gene Name
junctophilin 3
Gene Aliases
CAGL237, HDL2, JP-3, JP3, TNRC22
Chromosome Location
Chr.16: 87601835 - 87698156 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020655.3 NP_065706.2 2 - 3 1422 64
NR_073379.2 2 - 3 876 64
GenBank mRNA AB042636.1 - 2 - 3 1331 64
AK055486.1 - 1 - 2 233 64
AK295518.1 - 2 - 3 876 64
BC036533.2 - 2 - 3 1404 64

View Full Product Details