The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286747.1 | NP_001273676.1 | 2 - 3 | 428 | 120 |
GenBank mRNA | AK298608.1 | - | 2 - 3 | 254 | 120 |