The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252020.1 | NP_001238949.1 | 2 - 3 | 448 | 91 |
NM_001252024.1 | NP_001238953.1 | 3 - 4 | 236 | 91 | |
NM_001252030.1 | NP_001238959.1 | 2 - 3 | 150 | 91 | |
NM_002420.5 | NP_002411.3 | 2 - 3 | 150 | 91 | |
GenBank mRNA | AF071787.1 | - | 2 - 3 | 127 | 91 |
BC017849.1 | - | 2 - 3 | 145 | 91 | |
BC058286.1 | - | 2 - 3 | 154 | 91 | |
BU933033.1 | - | 2 - 3 | 153 | 91 | |
HM135790.1 | - | 3 - 4 | 231 | 91 | |
HM135791.1 | - | 2 - 3 | 448 | 91 |