The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252020.1 | NP_001238949.1 | 7 - 8 | 1329 | 118 |
NM_001252024.1 | NP_001238953.1 | 8 - 9 | 1117 | 118 | |
NM_002420.5 | NP_002411.3 | 7 - 8 | 1031 | 118 | |
GenBank mRNA | AB115498.1 | - | 4 - 5 | 688 | 118 |
AB115499.1 | - | 4 - 5 | 688 | 118 | |
AB115501.1 | - | 4 - 5 | 688 | 118 | |
AF071787.1 | - | 7 - 8 | 1008 | 118 | |
HM135790.1 | - | 8 - 9 | 1112 | 118 | |
HM135791.1 | - | 7 - 8 | 1329 | 118 |