Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR33
Entrez Gene ID
55339
Gene Name
WD repeat domain 33
Gene Aliases
NET14, WDC146
Chromosome Location
Chr.2: 127704234 - 127811187 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006623.2 NP_001006624.1 6 - 7 823 104
NM_018383.4 NP_060853.3 6 - 7 823 104
XM_005263697.2 XP_005263754.1 6 - 7 823 104
XM_011511436.1 XP_011509738.1 6 - 7 852 104
XM_017004435.1 XP_016859924.1 6 - 7 823 104
XM_017004436.1 XP_016859925.1 6 - 7 823 104
GenBank mRNA AB044749.1 - 6 - 7 783 104
BC068484.1 - 6 - 7 798 104

View Full Product Details