The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_033349.1 | 1 - 2 | 321 | 84 | |
GenBank mRNA | DA377448.1 | - | 1 - 2 | 266 | 84 |
DR005700.1 | - | 1 - 2 | 327 | 84 |