The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318360.1 | NP_001305289.1 | 18 - 19 | 2397 | 89 |
NM_003640.4 | NP_003631.2 | 18 - 19 | 2542 | 89 | |
XM_011519136.1 | XP_011517438.1 | 18 - 19 | 2334 | 89 | |
XM_011519137.1 | XP_011517439.1 | 18 - 19 | 2108 | 89 | |
XM_017015238.1 | XP_016870727.1 | 16 - 17 | 2113 | 89 | |
GenBank mRNA | AB210033.1 | - | 18 - 19 | 2304 | 89 |
AF044195.1 | - | 18 - 19 | 2321 | 89 | |
AF153419.2 | - | 18 - 19 | 2328 | 89 | |
AK001641.1 | - | 18 - 19 | 2326 | 89 | |
AK127237.1 | - | 10 - 11 | 1957 | 89 | |
AK225798.1 | - | 5 - 6 | 626 | 89 | |
AK289962.1 | - | 18 - 19 | 2334 | 89 | |
AK304742.1 | - | 18 - 19 | 2039 | 89 | |
BC033094.1 | - | 18 - 19 | 2104 | 89 | |
CR749385.1 | - | 11 - 12 | 1369 | 89 |