The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318360.1 | NP_001305289.1 | 25 - 26 | 3115 | 155 |
NM_003640.4 | NP_003631.2 | 25 - 26 | 3260 | 155 | |
XM_011519136.1 | XP_011517438.1 | 25 - 26 | 3052 | 155 | |
XM_011519137.1 | XP_011517439.1 | 25 - 26 | 2826 | 155 | |
XM_017015238.1 | XP_016870727.1 | 23 - 24 | 2831 | 155 | |
GenBank mRNA | AB210033.1 | - | 25 - 26 | 3022 | 155 |
AF044195.1 | - | 25 - 26 | 3039 | 155 | |
AF153419.2 | - | 25 - 26 | 3046 | 155 | |
AK001641.1 | - | 25 - 26 | 3044 | 155 | |
AK024223.1 | - | 5 - 6 | 503 | 155 | |
AK127237.1 | - | 17 - 18 | 2675 | 155 | |
AK225798.1 | - | 12 - 13 | 1344 | 155 | |
AK289962.1 | - | 25 - 26 | 3052 | 155 | |
AK304742.1 | - | 25 - 26 | 2757 | 155 | |
BC033094.1 | - | 25 - 26 | 2822 | 155 | |
CR749385.1 | - | 18 - 19 | 2087 | 155 |