The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308263.1 | NP_001295192.1 | 2 - 3 | 206 | 132 |
NM_003799.2 | NP_003790.1 | 2 - 3 | 206 | 132 | |
XM_011525749.1 | XP_011524051.1 | 2 - 3 | 219 | 132 | |
XM_011525750.2 | XP_011524052.1 | 2 - 3 | 234 | 132 | |
GenBank mRNA | AB007858.1 | - | 2 - 3 | 161 | 132 |
AB022604.1 | - | 1 - 2 | 132 | 132 | |
AB022605.1 | - | 1 - 2 | 132 | 132 | |
AF067791.1 | - | 2 - 3 | 161 | 132 | |
AK292561.1 | - | 2 - 3 | 163 | 132 | |
BC005290.1 | - | 2 - 3 | 170 | 132 |