Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRM3
Entrez Gene ID
2913
Gene Name
glutamate metabotropic receptor 3
Gene Aliases
GLUR3, GPRC1C, MGLUR3, mGlu3
Chromosome Location
Chr.7: 86643914 - 86864879 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000840.2 NP_000831.2 5 - 6 3672 76
XM_011516088.1 XP_011514390.1 4 - 5 1779 76
GenBank mRNA AK291406.1 - 5 - 6 3034 76
AK294178.1 - 3 - 4 1732 76
AK294701.1 - 4 - 5 2342 76
BC041407.1 - 5 - 6 3679 76
DQ315361.1 - 4 - 5 1677 76
DQ315362.1 - 4 - 5 2136 76
DQ315363.1 - 3 - 4 1280 76
EU432123.1 - 4 - 5 2573 76
X77748.1 - 5 - 6 2825 76

View Full Product Details