The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256482.1 | NP_001243411.1 | 8 - 9 | 1075 | 100 |
NM_001256483.1 | NP_001243412.1 | 7 - 8 | 1028 | 100 | |
NM_001256484.1 | NP_001243413.1 | 8 - 9 | 945 | 100 | |
NM_001979.5 | NP_001970.2 | 8 - 9 | 1113 | 100 | |
XM_017013199.1 | XP_016868688.1 | 7 - 8 | 841 | 100 | |
GenBank mRNA | AF233334.1 | - | 8 - 9 | 940 | 100 |
AF233335.1 | - | 8 - 9 | 940 | 100 | |
AF233336.1 | - | 8 - 9 | 940 | 100 | |
AK096089.1 | - | 7 - 8 | 879 | 100 | |
AK096770.1 | - | 8 - 9 | 911 | 100 | |
AK294065.1 | - | 8 - 9 | 918 | 100 | |
AK308041.1 | - | 8 - 9 | 918 | 100 | |
AK310507.1 | - | 8 - 9 | 990 | 100 | |
BC007708.2 | - | 8 - 9 | 940 | 100 | |
BC011628.2 | - | 8 - 9 | 971 | 100 | |
BC013874.2 | - | 8 - 9 | 934 | 100 | |
BT006885.1 | - | 8 - 9 | 906 | 100 | |
L05779.1 | - | 8 - 9 | 944 | 100 |