Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
STOML1
Entrez Gene ID
9399
Gene Name
stomatin like 1
Gene Aliases
SLP-1, STORP, hUNC-24
Chromosome Location
Chr.15: 73983218 - 73994622 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256672.1 NP_001243601.1 5 - 6 915 83
NM_001256674.1 NP_001243603.1 4 - 5 765 83
NM_001256676.1 NP_001243605.1 5 - 6 729 83
NM_001256677.1 NP_001243606.1 6 - 7 1046 83
NM_001324226.1 NP_001311155.1 4 - 5 1034 83
XM_006720773.3 XP_006720836.1 5 - 6 1820 83
GenBank mRNA AF074953.1 - 5 - 6 780 83
AK093689.1 - 1 - 2 1183 83
AK222665.1 - 5 - 6 798 83
AK300799.1 - 6 - 7 930 83
AL109664.1 - 4 - 5 595 83
AL109665.1 - 5 - 6 769 83
AL533460.3 - 5 - 6 706 83
BX464941.2 - 5 - 6 729 83

View Full Product Details