The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099855.1 | NP_001093325.1 | 12 - 13 | 1524 | 119 |
NM_001171876.1 | NP_001165347.1 | 12 - 13 | 1524 | 119 | |
NM_001171877.1 | NP_001165348.1 | 8 - 9 | 1389 | 119 | |
NM_001171878.1 | NP_001165349.1 | 9 - 10 | 1506 | 119 | |
NM_001171879.1 | NP_001165350.1 | 9 - 10 | 1506 | 119 | |
NM_005369.4 | NP_005360.3 | 9 - 10 | 1506 | 119 | |
XM_005262413.4 | XP_005262470.1 | 13 - 14 | 1672 | 119 | |
XM_011531339.2 | XP_011529641.1 | 13 - 14 | 1731 | 119 | |
XM_017029529.1 | XP_016885018.1 | 12 - 13 | 1493 | 119 | |
XM_017029530.1 | XP_016885019.1 | 12 - 13 | 1493 | 119 | |
XM_017029531.1 | XP_016885020.1 | 12 - 13 | 1426 | 119 | |
XM_017029532.1 | XP_016885021.1 | 9 - 10 | 1583 | 119 | |
GenBank mRNA | AB085901.1 | - | 9 - 10 | 1183 | 119 |
AB085902.1 | - | 9 - 10 | 1183 | 119 | |
AK296488.1 | - | 8 - 9 | 1389 | 119 | |
AK296529.1 | - | 12 - 13 | 1624 | 119 | |
AK302957.1 | - | 13 - 14 | 1472 | 119 | |
AK313900.1 | - | 9 - 10 | 1366 | 119 | |
AK316416.1 | - | 12 - 13 | 1512 | 119 | |
AL117234.1 | - | 11 - 12 | 1407 | 119 | |
X12556.1 | - | 9 - 10 | 1357 | 119 |