The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014509.4 | NP_055324.2 | 1 - 2 | 181 | 62 |
NR_104300.1 | 1 - 2 | 181 | 62 | ||
NR_104301.1 | 1 - 2 | 181 | 62 | ||
XM_017028738.1 | XP_016884227.1 | 1 - 2 | 170 | 62 | |
XM_017028739.1 | XP_016884228.1 | 1 - 2 | 168 | 62 | |
GenBank mRNA | AK294584.1 | - | 1 - 2 | 125 | 62 |
AK295223.1 | - | 1 - 2 | 87 | 62 | |
AL450314.1 | - | 1 - 2 | 124 | 62 | |
BC119650.2 | - | 1 - 2 | 65 | 62 | |
BC119651.1 | - | 1 - 2 | 65 | 62 | |
BC127635.1 | - | 1 - 2 | 66 | 62 | |
BC127636.1 | - | 1 - 2 | 66 | 62 | |
BI603374.1 | - | - | 189 | 62 | |
DC315058.1 | - | 1 - 2 | 158 | 62 |