Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAPK15
Entrez Gene ID
225689
Gene Name
mitogen-activated protein kinase 15
Gene Aliases
ERK7, ERK8
Chromosome Location
Chr.8: 143716305 - 143722463 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139021.2 NP_620590.2 11 - 12 1245 66
XM_011516925.2 XP_011515227.1 11 - 12 2083 66
XM_011516926.2 XP_011515228.1 11 - 12 1334 66
XM_011516927.2 XP_011515229.1 11 - 12 2026 66
XM_017013218.1 XP_016868707.1 10 - 11 1990 66
GenBank mRNA AY065978.1 - 11 - 12 1237 66
BC028034.1 - 9 - 10 1602 66

View Full Product Details