The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308263.1 | NP_001295192.1 | 11 - 12 | 1632 | 109 |
XM_017026061.1 | XP_016881550.1 | 10 - 11 | 1503 | 109 | |
GenBank mRNA | AB022605.1 | - | 10 - 11 | 1558 | 109 |
AK292561.1 | - | 11 - 12 | 1589 | 109 |