The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271895.1 | NP_001258824.1 | 14 - 15 | 1735 | 81 |
NM_001271896.1 | NP_001258825.1 | 12 - 13 | 1591 | 81 | |
NM_025150.4 | NP_079426.2 | 16 - 17 | 1981 | 81 | |
NR_073513.1 | 12 - 13 | 1380 | 81 | ||
NR_073514.1 | 15 - 16 | 1856 | 81 | ||
XM_006711555.1 | XP_006711618.1 | 15 - 16 | 1842 | 81 | |
XM_006711556.1 | XP_006711619.1 | 15 - 16 | 1797 | 81 | |
XM_017002394.1 | XP_016857883.1 | 14 - 15 | 1695 | 81 | |
XM_017002395.1 | XP_016857884.1 | 13 - 14 | 1596 | 81 | |
GenBank mRNA | AK022590.1 | - | 12 - 13 | 1541 | 81 |
AK222814.1 | - | 16 - 17 | 1922 | 81 | |
AK225368.1 | - | 14 - 15 | 1655 | 81 | |
AK315507.1 | - | 14 - 15 | 1681 | 81 | |
AL512732.1 | - | 5 - 6 | 445 | 81 | |
BC000541.2 | - | 16 - 17 | 1928 | 81 | |
BC007824.2 | - | 16 - 17 | 1869 | 81 | |
BC009997.2 | - | 16 - 17 | 1901 | 81 |