The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271895.1 | NP_001258824.1 | 10 - 11 | 1381 | 90 |
NM_001271896.1 | NP_001258825.1 | 8 - 9 | 1237 | 90 | |
NM_025150.4 | NP_079426.2 | 12 - 13 | 1627 | 90 | |
NR_073513.1 | 8 - 9 | 1026 | 90 | ||
NR_073514.1 | 11 - 12 | 1502 | 90 | ||
XM_006711555.1 | XP_006711618.1 | 12 - 13 | 1590 | 90 | |
XM_006711556.1 | XP_006711619.1 | 11 - 12 | 1443 | 90 | |
XM_017002394.1 | XP_016857883.1 | 11 - 12 | 1443 | 90 | |
XM_017002395.1 | XP_016857884.1 | 10 - 11 | 1344 | 90 | |
GenBank mRNA | AK022590.1 | - | 8 - 9 | 1187 | 90 |
AK222814.1 | - | 12 - 13 | 1568 | 90 | |
AK225368.1 | - | 10 - 11 | 1301 | 90 | |
AK315507.1 | - | 10 - 11 | 1327 | 90 | |
AL512732.1 | - | 1 - 2 | 91 | 90 | |
AY450558.1 | - | 8 - 9 | 956 | 90 | |
BC000541.2 | - | 12 - 13 | 1574 | 90 | |
BC007824.2 | - | 12 - 13 | 1515 | 90 | |
BC009997.2 | - | 12 - 13 | 1547 | 90 | |
BP308698.1 | - | 4 - 5 | 544 | 90 |