Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TARS2
Entrez Gene ID
80222
Gene Name
threonyl-tRNA synthetase 2, mitochondrial (putative)
Gene Aliases
COXPD21, TARSL1, thrRS
Chromosome Location
Chr.1: 150487364 - 150507609 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271895.1 NP_001258824.1 5 - 6 718 79
NM_025150.4 NP_079426.2 5 - 6 718 79
NR_073514.1 4 - 5 593 79
XM_006711555.1 XP_006711618.1 5 - 6 681 79
XM_006711556.1 XP_006711619.1 5 - 6 681 79
XM_017002394.1 XP_016857883.1 5 - 6 681 79
XM_017002395.1 XP_016857884.1 5 - 6 681 79
GenBank mRNA AK222814.1 - 5 - 6 659 79
AK225368.1 - 5 - 6 638 79
AK298773.1 - 4 - 5 546 79
AK315507.1 - 5 - 6 664 79
BC000541.2 - 5 - 6 665 79
BC007824.2 - 5 - 6 606 79
BC009997.2 - 5 - 6 638 79

View Full Product Details