The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206691.1 | NP_001193620.1 | 12 - 13 | 1405 | 67 |
NM_032491.5 | NP_115880.2 | 8 - 9 | 1071 | 67 | |
NM_213594.2 | NP_998759.1 | 12 - 13 | 1652 | 67 | |
GenBank mRNA | AB044245.1 | - | 12 - 13 | 1370 | 67 |
AB095365.1 | - | 12 - 13 | 1400 | 67 | |
AB095366.1 | - | 12 - 13 | 1652 | 67 | |
AB195784.1 | - | 2 - 3 | 245 | 67 | |
AB195785.1 | - | 2 - 3 | 167 | 67 | |
AF332192.1 | - | 8 - 9 | 67 | ||
AK315698.1 | - | 8 - 9 | 1051 | 67 | |
AL833921.1 | - | 5 - 6 | 475 | 67 | |
AY102009.1 | - | 12 - 13 | 1305 | 67 | |
BC028582.2 | - | 8 - 9 | 1074 | 67 | |
BC030644.1 | - | 8 - 9 | 1046 | 67 |