Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RFX4
Entrez Gene ID
5992
Gene Name
regulatory factor X4
Gene Aliases
NYD-SP10
Chromosome Location
Chr.12: 106582907 - 106762804 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206691.1 NP_001193620.1 13 - 14 1521 85
NM_032491.5 NP_115880.2 9 - 10 1187 85
NM_213594.2 NP_998759.1 13 - 14 1768 85
GenBank mRNA AB044245.1 - 13 - 14 1486 85
AB095365.1 - 13 - 14 1516 85
AB095366.1 - 13 - 14 1768 85
AB195784.1 - 3 - 4 361 85
AB195785.1 - 3 - 4 283 85
AF332192.1 - 9 - 10 1181 85
AK074913.1 - 10 - 11 1099 85
AK291445.1 - 13 - 14 1753 85
AK315698.1 - 9 - 10 1167 85
AL833921.1 - 6 - 7 591 85
AY102009.1 - 13 - 14 1421 85
BC028582.2 - 9 - 10 1190 85
BC030644.1 - 9 - 10 1162 85

View Full Product Details