Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RFX4
Entrez Gene ID
5992
Gene Name
regulatory factor X4
Gene Aliases
NYD-SP10
Chromosome Location
Chr.12: 106582907 - 106762804 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206691.1 NP_001193620.1 2 - 3 290 112
NM_213594.2 NP_998759.1 2 - 3 537 112
GenBank mRNA AB044245.1 - 2 - 3 255 112
AB095365.1 - 2 - 3 285 112
AB095366.1 - 2 - 3 537 112
AK291445.1 - 2 - 3 522 112
AK302837.1 - 2 - 3 261 112
AY102009.1 - 2 - 3 190 112
DB445991.1 - 2 - 4 290 112

View Full Product Details