The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081559.2 | NP_001075028.1 | 6 - 7 | 715 | 97 |
NM_001318161.1 | NP_001305090.1 | 5 - 6 | 621 | 97 | |
NM_001318162.1 | NP_001305091.1 | 6 - 7 | 690 | 97 | |
XM_011515756.2 | XP_011514058.1 | 6 - 7 | 733 | 97 | |
GenBank mRNA | AK301745.1 | - | 6 - 7 | 645 | 97 |
BC003101.1 | - | 6 - 7 | 630 | 97 | |
BE388179.1 | - | 4 - 5 | 390 | 97 | |
BQ052009.1 | - | 6 - 7 | 677 | 97 |