The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145246.4 | NP_660289.2 | 7 - 8 | 660 | 151 |
XM_005269515.4 | XP_005269572.1 | 8 - 9 | 757 | 151 | |
XM_005269516.4 | XP_005269573.1 | 7 - 8 | 852 | 151 | |
XM_011539259.2 | XP_011537561.1 | 7 - 8 | 625 | 151 | |
XM_011539260.2 | XP_011537562.1 | 8 - 9 | 715 | 151 | |
XM_017015656.1 | XP_016871145.1 | 7 - 8 | 625 | 151 | |
XM_017015657.1 | XP_016871146.1 | 7 - 8 | 660 | 151 | |
XM_017015658.1 | XP_016871147.1 | 8 - 9 | 760 | 151 | |
XM_017015659.1 | XP_016871148.1 | 7 - 8 | 852 | 151 | |
XM_017015660.1 | XP_016871149.1 | 8 - 9 | 717 | 151 | |
XM_017015661.1 | XP_016871150.1 | 8 - 9 | 774 | 151 | |
XM_017015662.1 | XP_016871151.1 | 7 - 8 | 625 | 151 | |
GenBank mRNA | AJ431721.1 | - | 7 - 8 | 656 | 151 |
AJ431722.1 | - | 7 - 8 | 656 | 151 | |
AJ431723.1 | - | 7 - 8 | 656 | 151 | |
AJ431724.1 | - | 7 - 8 | 656 | 151 | |
AJ431725.1 | - | 7 - 8 | 656 | 151 | |
AK090955.1 | - | 7 - 8 | 660 | 151 | |
BC018007.1 | - | 7 - 8 | 529 | 151 |