Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EXOC6
Entrez Gene ID
54536
Gene Name
exocyst complex component 6
Gene Aliases
EXOC6A, SEC15, SEC15L, SEC15L1, SEC15L3, Sec15p
Chromosome Location
Chr.10: 92826831 - 93059494 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013848.3 NP_001013870.1 19 - 20 2080 119
NM_001319194.1 NP_001306123.1 19 - 20 2146 119
NM_001319195.1 NP_001306124.1 18 - 19 2031 119
NM_001319200.1 NP_001306129.1 14 - 15 1725 119
NM_019053.5 NP_061926.3 18 - 19 2034 119
XM_017016344.1 XP_016871833.1 19 - 20 2138 119
XM_017016345.1 XP_016871834.1 18 - 19 2115 119
XM_017016346.1 XP_016871835.1 18 - 19 2112 119
XM_017016347.1 XP_016871836.1 18 - 19 2318 119
XM_017016348.1 XP_016871837.1 19 - 20 2141 119
GenBank mRNA AK002113.1 - 10 - 11 1032 119
AK123268.1 - 5 - 6 1389 119
AK128190.1 - 19 - 20 2080 119
AK293857.1 - 18 - 19 1997 119
AK300285.1 - 19 - 20 2146 119
AK315544.1 - 18 - 19 1969 119
AL137438.1 - 13 - 14 1331 119
AL832501.1 - 18 - 19 1975 119
BC028395.2 - 18 - 19 1969 119
EF571007.1 - 14 - 15 1666 119

View Full Product Details