The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013848.3 | NP_001013870.1 | 3 - 4 | 399 | 130 |
NM_001319194.1 | NP_001306123.1 | 3 - 4 | 465 | 130 | |
NM_001319195.1 | NP_001306124.1 | 2 - 3 | 353 | 130 | |
NM_001319200.1 | NP_001306129.1 | 2 - 3 | 353 | 130 | |
NM_019053.5 | NP_061926.3 | 2 - 3 | 353 | 130 | |
XM_017016344.1 | XP_016871833.1 | 3 - 4 | 460 | 130 | |
XM_017016345.1 | XP_016871834.1 | 2 - 3 | 434 | 130 | |
XM_017016346.1 | XP_016871835.1 | 2 - 3 | 434 | 130 | |
XM_017016347.1 | XP_016871836.1 | 2 - 3 | 637 | 130 | |
XM_017016348.1 | XP_016871837.1 | 3 - 4 | 460 | 130 | |
GenBank mRNA | AF220217.1 | - | 2 - 3 | 336 | 130 |
AK128190.1 | - | 3 - 4 | 399 | 130 | |
AK293857.1 | - | 2 - 3 | 316 | 130 | |
AK300285.1 | - | 3 - 4 | 465 | 130 | |
AK315544.1 | - | 2 - 3 | 288 | 130 | |
AL832501.1 | - | 2 - 3 | 297 | 130 | |
BC028395.2 | - | 2 - 3 | 288 | 130 | |
BM148225.1 | - | 2 - 3 | 323 | 130 | |
CB991016.1 | - | 3 - 4 | 395 | 130 | |
DA187840.1 | - | 2 - 3 | 340 | 130 | |
DA333847.1 | - | 2 - 3 | 354 | 130 | |
DB060830.1 | - | 2 - 3 | 437 | 130 | |
EF571007.1 | - | 2 - 3 | 294 | 130 |