Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EXOC6
Entrez Gene ID
54536
Gene Name
exocyst complex component 6
Gene Aliases
EXOC6A, SEC15, SEC15L, SEC15L1, SEC15L3, Sec15p
Chromosome Location
Chr.10: 92826831 - 93059494 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013848.3 NP_001013870.1 7 - 8 789 148
NM_001319194.1 NP_001306123.1 7 - 8 855 148
NM_001319195.1 NP_001306124.1 6 - 7 743 148
NM_001319200.1 NP_001306129.1 6 - 7 743 148
NM_019053.5 NP_061926.3 6 - 7 743 148
XM_017016344.1 XP_016871833.1 7 - 8 850 148
XM_017016345.1 XP_016871834.1 6 - 7 824 148
XM_017016346.1 XP_016871835.1 6 - 7 824 148
XM_017016347.1 XP_016871836.1 6 - 7 1027 148
XM_017016348.1 XP_016871837.1 7 - 8 850 148
GenBank mRNA AK128190.1 - 7 - 8 789 148
AK293857.1 - 6 - 7 706 148
AK300285.1 - 7 - 8 855 148
AK315544.1 - 6 - 7 678 148
AL832501.1 - 6 - 7 687 148
BC028395.2 - 6 - 7 678 148
EF571007.1 - 6 - 7 684 148

View Full Product Details