The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005479.3 | NP_005470.2 | 1 - 1 | 2244 | 78 |
GenBank mRNA | U58975.2 | - | 1 - 1 | 2243 | 78 |