The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021942.5 | NP_068761.4 | 11 - 12 | 1405 | 110 |
NM_199053.2 | NP_951008.1 | 11 - 12 | 1405 | 110 | |
XM_017008537.1 | XP_016864026.1 | 11 - 12 | 1405 | 110 | |
XM_017008538.1 | XP_016864027.1 | 11 - 12 | 1405 | 110 | |
GenBank mRNA | AK091146.1 | - | 6 - 7 | 629 | 110 |
AK096345.1 | - | 11 - 12 | 1379 | 110 | |
AK315057.1 | - | 6 - 7 | 611 | 110 | |
AL136752.1 | - | 3 - 4 | 314 | 110 | |
AL833451.1 | - | 10 - 11 | 1213 | 110 | |
AL833571.1 | - | 11 - 12 | 1384 | 110 | |
BC051724.1 | - | 11 - 12 | 1349 | 110 | |
BX538044.1 | - | 11 - 12 | 1359 | 110 | |
BX647127.1 | - | 11 - 12 | 1376 | 110 | |
BX648499.1 | - | 11 - 12 | 1380 | 110 | |
CR533553.1 | - | 3 - 4 | 249 | 110 |