The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021942.5 | NP_068761.4 | 18 - 19 | 2098 | 121 |
NM_199053.2 | NP_951008.1 | 18 - 19 | 2098 | 121 | |
XM_017008537.1 | XP_016864026.1 | 18 - 19 | 2098 | 121 | |
XM_017008538.1 | XP_016864027.1 | 18 - 19 | 2098 | 121 | |
XM_017008539.1 | XP_016864028.1 | 16 - 17 | 1749 | 121 | |
GenBank mRNA | AK026992.1 | - | 4 - 5 | 490 | 121 |
AK091146.1 | - | 13 - 14 | 1322 | 121 | |
AK096345.1 | - | 18 - 19 | 2072 | 121 | |
AK315057.1 | - | 13 - 14 | 1304 | 121 | |
AL136752.1 | - | 10 - 11 | 1007 | 121 | |
AL833451.1 | - | 17 - 18 | 1906 | 121 | |
AL833571.1 | - | 18 - 19 | 2077 | 121 | |
BC051724.1 | - | 18 - 19 | 2042 | 121 | |
BX538044.1 | - | 18 - 19 | 2052 | 121 | |
BX647127.1 | - | 18 - 19 | 2069 | 121 | |
BX648499.1 | - | 18 - 19 | 2073 | 121 | |
CR533553.1 | - | 10 - 11 | 942 | 121 |