Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COQ2
Entrez Gene ID
27235
Gene Name
coenzyme Q2, polyprenyltransferase
Gene Aliases
CL640, COQ10D1, MSA1, PHB:PPT
Chromosome Location
Chr.4: 83138568 - 83285129 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015697.7 NP_056512.5 2 - 3 572 70
XM_011531855.2 XP_011530157.1 2 - 3 783 70
XM_011531857.2 XP_011530159.1 2 - 3 781 70
XM_011531859.2 XP_011530161.1 2 - 3 780 70
XM_011531860.2 XP_011530162.1 2 - 3 780 70
XM_011531862.2 XP_011530164.1 2 - 3 779 70
XM_011531863.2 XP_011530165.1 2 - 3 776 70
XM_011531866.2 XP_011530168.1 2 - 3 772 70
XM_011531867.2 XP_011530169.1 2 - 3 695 70
XM_017008031.1 XP_016863520.1 2 - 3 695 70
GenBank mRNA AF091086.1 - 1 - 2 122 70
AJ621061.3 - 2 - 3 572 70
AK129583.1 - 2 - 3 328 70
AK300972.1 - 2 - 3 453 70
BC008804.2 - 2 - 3 429 70
BC020728.1 - 2 - 3 469 70
BG699931.1 - 2 - 3 458 70

View Full Product Details