Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRIMA1
Entrez Gene ID
145270
Gene Name
proline rich membrane anchor 1
Gene Aliases
PRIMA
Chromosome Location
Chr.14: 93718298 - 93789029 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AY225517.1 - 4 - 5 573 83

View Full Product Details