Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LAMA4
Entrez Gene ID
3910
Gene Name
laminin subunit alpha 4
Gene Aliases
CMD1JJ, LAMA3, LAMA4*-1
Chromosome Location
Chr.6: 112107931 - 112254722 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105206.2 NP_001098676.2 35 - 36 5375 106
NM_001105207.2 NP_001098677.2 35 - 36 5371 106
NM_002290.4 NP_002281.3 35 - 36 5354 106
XM_005266983.3 XP_005267040.2 35 - 36 5248 106
XM_005266984.3 XP_005267041.2 35 - 36 5245 106
XM_017010854.1 XP_016866343.1 35 - 36 5224 106
GenBank mRNA AB210027.1 - 35 - 36 5227 106
BC066552.1 - 35 - 36 5367 106
S78569.1 - 35 - 36 5146 106
X70904.1 - 5 - 6 750 106
X76939.1 - 29 - 30 4251 106
X91171.1 - 35 - 36 5239 106

View Full Product Details