Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
QTRT1
Entrez Gene ID
81890
Gene Name
queuine tRNA-ribosyltransferase catalytic subunit 1
Gene Aliases
FP3235, TGT, TGUT
Chromosome Location
Chr.19: 10701436 - 10713367 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031209.2 NP_112486.1 9 - 10 1090 74
GenBank mRNA AF302783.1 - 9 - 10 1071 74
AF302784.1 - 9 - 10 995 74
AF302785.1 - - 1149 74
AF370379.1 - 8 - 9 1267 74
AK055216.1 - 3 - 4 1793 74
AK294166.1 - 8 - 9 1359 74
AK314308.1 - 9 - 10 1075 74
BC015350.1 - 9 - 10 1049 74

View Full Product Details