Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KL
Entrez Gene ID
9365
Gene Name
klotho
Gene Aliases
-
Chromosome Location
Chr.13: 33016063 - 33066145 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004795.3 NP_004786.2 4 - 5 2710 114
XM_006719895.2 XP_006719958.1 4 - 5 2134 114
GenBank mRNA AB005142.1 - 4 - 5 2710 114

View Full Product Details