Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NLRP11
Entrez Gene ID
204801
Gene Name
NLR family pyrin domain containing 11
Gene Aliases
CLR19.6, NALP11, NOD17, PAN10, PYPAF6, PYPAF7
Chromosome Location
Chr.19: 55785397 - 55836762 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297743.1 NP_001284672.1 4 - 5 2034 60
NM_145007.3 NP_659444.2 7 - 8 2884 60
GenBank mRNA AK090621.1 - 2 - 3 922 60
AK129784.1 - 2 - 3 949 60
AY095145.1 - 5 - 6 2272 60
AY154466.1 - 4 - 5 2173 60
AY168967.1 - 5 - 6 2261 60
AY168968.1 - 4 - 5 1933 60
AY226382.1 - 4 - 5 2173 60
BC034730.1 - 5 - 6 2261 60

View Full Product Details