Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NLRP11
Entrez Gene ID
204801
Gene Name
NLR family pyrin domain containing 11
Gene Aliases
CLR19.6, NALP11, NOD17, PAN10, PYPAF6, PYPAF7
Chromosome Location
Chr.19: 55785397 - 55836762 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297743.1 NP_001284672.1 7 - 8 2548 71
NM_145007.3 NP_659444.2 10 - 11 3398 71
GenBank mRNA AK090621.1 - 5 - 6 1436 71
AK129784.1 - 5 - 6 1463 71
AY095145.1 - 8 - 9 2786 71
AY154466.1 - 7 - 8 2687 71
AY168967.1 - 8 - 9 2775 71
AY168968.1 - 7 - 8 2447 71
AY226382.1 - 7 - 8 2687 71
BC034730.1 - 8 - 9 2775 71

View Full Product Details