The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318467.1 | NP_001305396.1 | 1 - 2 | 397 | 58 |
NM_031291.3 | NP_112581.1 | 1 - 2 | 397 | 58 | |
XM_011532298.1 | XP_011530600.1 | 1 - 2 | 400 | 58 | |
GenBank mRNA | AJ863129.1 | - | 1 - 2 | 229 | 58 |
AL136857.1 | - | 1 - 2 | 356 | 58 | |
AY550240.1 | - | 1 - 2 | 348 | 58 | |
BC022032.2 | - | 1 - 2 | 348 | 58 | |
BG773445.1 | - | 1 - 2 | 380 | 58 | |
BM563946.1 | - | 1 - 2 | 389 | 58 | |
BQ438347.1 | - | 1 - 2 | 400 | 58 |