The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318467.1 | NP_001305396.1 | 4 - 5 | 802 | 108 |
NM_031291.3 | NP_112581.1 | 4 - 5 | 802 | 108 | |
GenBank mRNA | AJ863129.1 | - | 4 - 5 | 634 | 108 |
AL136857.1 | - | 4 - 5 | 761 | 108 | |
AY550240.1 | - | 4 - 5 | 753 | 108 | |
BC022032.2 | - | 4 - 5 | 753 | 108 | |
BM563946.1 | - | 4 - 5 | 794 | 108 |