The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001029875.2 | NP_001025046.1 | 1 - 2 | 847 | 75 |
XM_005248502.3 | XP_005248559.1 | 1 - 2 | 2229 | 75 | |
GenBank mRNA | AK296443.1 | - | 1 - 2 | 495 | 75 |
BC140707.1 | - | 1 - 2 | 193 | 75 | |
BX640900.1 | - | 1 - 2 | 85 | 75 |