The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001029875.2 | NP_001025046.1 | 3 - 4 | 1143 | 136 |
NM_001271890.1 | NP_001258819.1 | 1 - 2 | 257 | 136 | |
NM_001271891.1 | NP_001258820.1 | 1 - 2 | 257 | 136 | |
NR_073511.1 | 1 - 2 | 257 | 136 | ||
XM_005248502.3 | XP_005248559.1 | 3 - 4 | 2525 | 136 | |
GenBank mRNA | AK294808.1 | - | 2 - 3 | 248 | 136 |
AK296443.1 | - | 3 - 4 | 791 | 136 | |
AK296494.1 | - | 1 - 2 | 225 | 136 | |
BC140707.1 | - | 3 - 4 | 489 | 136 | |
BX640900.1 | - | 3 - 4 | 381 | 136 | |
DA211241.1 | - | 1 - 2 | 257 | 136 | |
DA417118.1 | - | 1 - 2 | 225 | 136 | |
DC342795.1 | - | 1 - 2 | 210 | 136 |