Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C11orf54
Entrez Gene ID
28970
Gene Name
chromosome 11 open reading frame 54
Gene Aliases
PTD012, PTOD012
Chromosome Location
Chr.11: 93741594 - 93765323 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286068.1 NP_001272997.1 1 - 2 128 61
XM_017017616.1 XP_016873105.1 1 - 2 125 61
XM_017017618.1 XP_016873107.1 1 - 2 124 61
GenBank mRNA AF092133.1 - 1 - 2 28 61
AK225977.1 - 1 - 2 63 61

View Full Product Details